siRNA Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129758.1 | NP_001123230.1 | 3 | 698 | ||
| NM_022357.3 | NP_071752.3 | 3 | 698 | |||
| GenBank mRNA | AJ291679.1 | - | 3 | 612 | ||
| AK057401.1 | - | 3 | 698 | |||
| AY358390.1 | - | 3 | 621 | |||
| BC037243.1 | - | 3 | 610 | |||
| BC057789.1 | - | 3 | 639 | 5297958 | ||

DPEP3
Genomic Map