siRNA Details
| Gene Symbol: NUP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024057.3 | NP_076962.2 | 3 | 323 | ||
| GenBank mRNA | AF514994.1 | - | 2 | 177 | ||
| AK026271.1 | - | 2 | 158 | |||
| AK296733.1 | - | 3 | 281 | |||
| AK310619.1 | - | 3 | 283 | |||
| BC000861.2 | - | 2 | 241 | 3459639 | ||
| DC420994.1 | - | 3 | 324 | |||

NUP37
Genomic Map