This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF557 SIRNA Assays ›
Gene Symbol:
ZNF557
Gene Name:
zinc finger protein 557
Gene Aliases:
-
Chromosome Location:
Chr.19: 7069460 - 7087968 on Build GRCh38
UniGene ID:
Hs.591380
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS128358
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF557
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044387.1 NP_001037852.1 8 914
NM_001044388.1 NP_001037853.1 8 905
NM_024341.2 NP_077317.2 8 926
GenBank mRNA AK095524.1 - 8 659
AK297283.1 - 8 683
AK315405.1 - 8 943
BC003020.1 - 8 676 2822966
BC031208.1 - 8 683
BC058040.1 - 8 655

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline