This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRC8E SIRNA Assays ›
Gene Symbol:
LRRC8E
Gene Name:
leucine rich repeat containing 8 family member E
Gene Aliases:
-
Chromosome Location:
Chr.19: 7888505 - 7902023 on Build GRCh38
UniGene ID:
Hs.501511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS129386
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC8E
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001268284.2 NP_001255213.1 4 1851
NM_001268285.2 NP_001255214.1 2 1565
NM_025061.5 NP_079337.2 3 1708
XM_011528319.2 XP_011526621.1 3 2436
GenBank mRNA AK027073.1 - 1 656
AK091134.1 - 3 1708
AL834474.1 - 1 1087
BC022216.1 - 1 996
BC070089.1 - 3 1708 30346745
BC108252.1 - 1 817
BX538180.1 - 3 1698

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of anion transport
anion transmembrane transport
volume-sensitive anion channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jrd88:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0