siRNA Details
| Gene Symbol: MARVELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031484.3 | NP_113672.1 | 1 | 634 | ||
| GenBank mRNA | AF325259.1 | - | 1 | 616 | ||
| AK025996.1 | - | 1 | 319 | |||
| BC004995.1 | - | 1 | 618 | |||
| BC062544.1 | - | 1 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARVELD1| Gene Symbol: MARVELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031484.3 | NP_113672.1 | 1 | 634 | ||
| GenBank mRNA | AF325259.1 | - | 1 | 616 | ||
| AK025996.1 | - | 1 | 319 | |||
| BC004995.1 | - | 1 | 618 | |||
| BC062544.1 | - | 1 | 620 | |||