This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NICN1 SIRNA Assays ›
Gene Symbol:
NICN1
Gene Name:
nicolin 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 49422333 - 49429324 on Build GRCh38
UniGene ID:
Hs.191460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS130848
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NICN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032316.3 NP_115692.1 6 711
GenBank mRNA AF538150.1 - 5 975
AJ299740.1 - 6 683
AK094248.1 - 6 651
AK301846.1 - 5 600
AK313073.1 - 6 708
BC005050.1 - 6 628 2820022
BC050653.1 - 6 675 6015080

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4m7g7:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline