This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ACSS1 SIRNA Assays ›
Gene Symbol:
ACSS1
Gene Name:
acyl-CoA synthetase short-chain family member 1
Gene Aliases:
ACAS2L, ACECS1, AceCS2L
Chromosome Location:
Chr.20: 25006230 - 25058182 on Build GRCh38
UniGene ID:
Hs.529353
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS131078
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ACSS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252675.1 NP_001239604.1 4, 5 898
NM_001252676.1 NP_001239605.1 3, 4 725
NM_001252677.1 NP_001239606.1 4, 5 898
NM_032501.3 NP_115890.2 4, 5 898
XM_011529388.1 XP_011527690.1 3, 4 683
GenBank mRNA AK125058.1 - 2, 3 2101
AK127566.1 - 4, 5 844
AK296306.1 - 3, 4 722
AK314440.1 - 4, 5 821
BC039261.1 - 4, 5 857 5298849
BC044588.1 - 4, 5 823 5260496
BC055008.1 - 4, 5 823 4838976
BC114612.1 - 4, 5 689

Back To Top

More Information


Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

ethanol oxidation
acetyl-CoA biosynthetic process
acetate biosynthetic process
acetyl-CoA biosynthetic process from acetate
propionate biosynthetic process
acetate-CoA ligase activity
protein binding
ATP binding
AMP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5xfzg:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline