siRNA Details
| Gene Symbol: ZNF607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172677.1 | NP_001166148.1 | 5 | 531 | ||
| NM_032689.4 | NP_116078.4 | 5 | 882 | |||
| XM_006723435.3 | XP_006723498.1 | 5 | 1136 | |||
| XM_006723436.2 | XP_006723499.1 | 5 | 846 | |||
| XM_006723437.2 | XP_006723500.1 | 4 | 750 | |||
| XM_006723440.1 | XP_006723503.1 | 2 | 573 | |||
| XM_011527414.2 | XP_011525716.1 | 2 | 827 | |||
| GenBank mRNA | AK131557.1 | - | 5 | 882 | ||
| AK131559.1 | - | 5 | 531 | |||

ZNF607
Genomic Map