This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STPG1 SIRNA Assays ›
Gene Symbol:
STPG1
Gene Name:
sperm tail PG-rich repeat containing 1
Gene Aliases:
C1orf201, MAPO2
Chromosome Location:
Chr.1: 24356999 - 24415533 on Build GRCh38
UniGene ID:
Hs.403187
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS132000
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STPG1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199012.1 NP_001185941.1 8 1105
NM_001199013.1 NP_001185942.1 8 1017
NM_001199014.1 NP_001185943.1 9 1161
NM_178122.4 NP_835223.1 7 898
XM_006711025.2 XP_006711088.1 9 1123
XM_011542403.2 XP_011540705.1 7 1422
XM_011542404.1 XP_011540706.1 9 1236
XM_017002768.1 XP_016858257.1 9 1232
XM_017002769.1 XP_016858258.1 10 1380
XM_017002770.1 XP_016858259.1 11 1495
GenBank mRNA BC017650.1 - 4 424
BC035061.1 - 9 1134 5262741
BC047705.1 - 6 803 5764388
BC063891.1 - 8 1049 30390664

Back To Top

More Information


Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s)

apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-rsggf:80/100.66.72.30:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0