siRNA Details
| Gene Symbol: OSBPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145798.2 | NP_665741.1 | 17, 18 | 2009 | ||
| XM_017024122.1 | XP_016879611.1 | 17, 18 | 2031 | |||
| XM_017024123.1 | XP_016879612.1 | 17, 18 | 2149 | |||
| XM_017024124.1 | XP_016879613.1 | 16, 17 | 1996 | |||
| XM_017024125.1 | XP_016879614.1 | 7, 8 | 932 | |||
| GenBank mRNA | AB208886.1 | - | 5, 6 | 2856 | ||
| AF323729.1 | - | 17, 18 | 1998 | |||
| AF392446.1 | - | 17, 18 | 2062 | |||
| BC065482.1 | - | 17, 18 | 1981 | 6081784 | ||

OSBPL7
Genomic Map