siRNA Details
| Gene Symbol: OSBPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145798.2 | NP_665741.1 | 23 | 2654 | ||
| XM_017024122.1 | XP_016879611.1 | 22 | 2814 | |||
| XM_017024123.1 | XP_016879612.1 | 22 | 2932 | |||
| XM_017024124.1 | XP_016879613.1 | 21 | 2779 | |||
| XM_017024125.1 | XP_016879614.1 | 12 | 1715 | |||
| GenBank mRNA | AB208886.1 | - | 11 | 3501 | ||
| AF323729.1 | - | 23 | 2643 | |||
| AF392446.1 | - | 23 | 2707 | |||
| BC028010.1 | - | 15 | 1842 | |||
| BC065482.1 | - | 23 | 2626 | 6081784 | ||

OSBPL7
Genomic Map