This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CLRN3 SIRNA Assays ›
Gene Symbol:
CLRN3
Gene Name:
clarin 3
Gene Aliases:
TMEM12, USH3AL1
Chromosome Location:
Chr.10: 127877841 - 127892947 on Build GRCh38
UniGene ID:
Hs.242014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS133379
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CLRN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152311.3 NP_689524.1 1 350
XM_011539274.2 XP_011537576.1 1 349
GenBank mRNA BC029478.1 - 1 350 4733535
BM830163.1 - 1 103

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0