siRNA Details
Gene Symbol: C11orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144663.1 | NP_653264.1 | 3 | 424 | ||
GenBank mRNA | AF439154.1 | - | 3 | 424 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C11orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144663.1 | NP_653264.1 | 3 | 424 | ||
GenBank mRNA | AF439154.1 | - | 3 | 424 |