This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF569 SIRNA Assays ›
Gene Symbol:
ZNF569
Gene Name:
zinc finger protein 569
Gene Aliases:
ZAP1, ZNF
Chromosome Location:
Chr.19: 37411146 - 37469268 on Build GRCh38
UniGene ID:
Hs.511848
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS135449
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF569
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152484.2 NP_689697.2 5 764
XM_006723046.2 XP_006723109.1 7 1091
XM_006723047.3 XP_006723110.1 6 1014
XM_006723048.3 XP_006723111.1 6 938
XM_011526538.2 XP_011524840.1 6 853
XM_011526539.2 XP_011524841.1 4 347
XM_017026376.1 XP_016881865.1 5 599
XM_017026377.1 XP_016881866.1 5 678
XM_017026378.1 XP_016881867.1 6 716
GenBank mRNA AK122846.1 - 2 85
AK289987.1 - 5 537
AL833408.1 - 5 765
AY835704.1 - 5 764
BC059407.1 - 4 392

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline