siRNA Details
| Gene Symbol: BROX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288579.1 | NP_001275508.1 | 12 | 1499 | ||
| NM_001288580.1 | NP_001275509.1 | 12 | 1205 | |||
| NM_001288581.1 | NP_001275510.1 | 12 | 1141 | |||
| NM_144695.3 | NP_653296.2 | 13 | 1595 | |||
| XM_005273065.2 | XP_005273122.2 | 13 | 1231 | |||
| XM_005273069.4 | XP_005273126.1 | 12 | 1467 | |||
| XM_006711173.3 | XP_006711236.1 | 14 | 1664 | |||
| XM_011509212.2 | XP_011507514.1 | 12 | 1558 | |||
| XM_011509213.2 | XP_011507515.1 | 12 | 1539 | |||
| XM_011509214.2 | XP_011507516.1 | 14 | 1464 | |||
| XM_017000374.1 | XP_016855863.1 | 15 | 1789 | |||
| XM_017000375.1 | XP_016855864.1 | 11 | 1444 | |||
| XM_017000376.1 | XP_016855865.1 | 13 | 1592 | |||
| GenBank mRNA | AB276123.1 | - | 12 | 1189 | ||
| AI243754.1 | - | 1 | 216 | |||
| AI911339.1 | - | 1 | 97 | |||
| AK056983.1 | - | 13 | 1584 | |||
| AK293974.1 | - | 12 | 1131 | |||
| AK304445.1 | - | 12 | 1086 | |||
| AK315930.1 | - | 12 | 1135 | |||
| BC113635.1 | - | 13 | 1332 | 8322687 | ||
| BC113637.1 | - | 13 | 1332 | 8322689 | ||
| BC143811.1 | - | 12 | 1236 | 9052326 | ||

BROX
Genomic Map