siRNA Details
| Gene Symbol: CFAP206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031743.2 | NP_001026913.1 | 5 | 578 | ||
| GenBank mRNA | AK093830.1 | - | 5 | 537 | ||
| BC035083.2 | - | 5 | 547 | 5260875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP206| Gene Symbol: CFAP206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031743.2 | NP_001026913.1 | 5 | 578 | ||
| GenBank mRNA | AK093830.1 | - | 5 | 537 | ||
| BC035083.2 | - | 5 | 547 | 5260875 | ||