siRNA Details
| Gene Symbol: C8orf37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177965.3 | NP_808880.1 | 6 | 594 | ||
| XM_005250799.3 | XP_005250856.2 | 5 | 1866 | |||
| GenBank mRNA | AK055162.1 | - | 6 | 569 | ||
| BC036855.2 | - | 6 | 559 | |||
| DB205618.1 | - | 6 | 559 | |||
| GQ240139.1 | - | 6 | 569 | |||

C8orf37
Genomic Map