This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FSIP1 SIRNA Assays ›
Gene Symbol:
FSIP1
Gene Name:
fibrous sheath interacting protein 1
Gene Aliases:
HSD10
Chromosome Location:
Chr.15: 39598643 - 39782838 on Build GRCh38
UniGene ID:
Hs.129598
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS136277
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FSIP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324338.1 NP_001311267.1 10 1258
NM_152597.4 NP_689810.3 10 1355
XM_011521305.2 XP_011519607.1 10 1231
XM_011521306.1 XP_011519608.1 10 1644
XM_011521307.2 XP_011519609.1 10 1231
XM_011521309.2 XP_011519611.1 10 1231
XM_011521311.2 XP_011519613.1 6 1915
XM_017021972.1 XP_016877461.1 6 708
GenBank mRNA AK093308.1 - 10 1337
AY260140.1 - 9 1152
BC045191.2 - 10 1347 5297527

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-qhjbl:80/100.66.79.187:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline