This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPTSSA SIRNA Assays ›
Gene Symbol:
SPTSSA
Gene Name:
serine palmitoyltransferase small subunit A
Gene Aliases:
C14orf147, SSSPTA
Chromosome Location:
Chr.14: 34432938 - 34462262 on Build GRCh38
UniGene ID:
Hs.740577
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS136701
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPTSSA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138288.3 NP_612145.2 2 202
GenBank mRNA AK001993.1 - 2 183
AK315409.1 - 2 180
AL080066.1 - 2 183
BC016805.1 - 2 184 4075637
BC016808.1 - 2 184 4077762
BC021701.1 - 2 183 4069530
BC068480.1 - 2 296 5294549

Back To Top

More Information


Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

sphingolipid biosynthetic process
ceramide biosynthetic process
serine C-palmitoyltransferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline