siRNA Details
| Gene Symbol: TMEM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178505.7 | NP_848600.2 | 6 | 1182 | ||
| NR_134507.1 | 6 | 1182 | ||||
| GenBank mRNA | AK124367.1 | - | 2 | 1173 | ||
| AK125611.1 | - | 6 | 970 | |||
| CR749606.1 | - | 6 | 1182 | |||

TMEM26
Genomic Map