siRNA Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152773.4 | NP_689986.2 | 3 | 428 | ||
| GenBank mRNA | BC021177.2 | - | 3 | 408 | 4830500 | |
| BM548106.1 | - | 3 | 453 | |||
| CK818899.1 | - | 3 | 422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCTEX1D2| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152773.4 | NP_689986.2 | 3 | 428 | ||
| GenBank mRNA | BC021177.2 | - | 3 | 408 | 4830500 | |
| BM548106.1 | - | 3 | 453 | |||
| CK818899.1 | - | 3 | 422 | |||