This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CH507-42P11.6 SIRNA Assays ›
Gene Symbol:
CH507-42P11.6 LINC00319
Gene Name:
uncharacterized CH507-42P11.6
long intergenic non-protein coding RNA 319
Gene Aliases:
-
C21orf125, NCRNA00319, PRED49
Chromosome Location:
Chr.21: 6084373 - 6088242 on Build GRCh38
Chromosome Location:
Chr.21: 43450024 - 43453893 on Build GRCh38
UniGene ID:
-
Hs.146127
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS138635
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CH507-42P11.6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006723919.2 XP_006723982.1 1 360
XM_011546086.1 XP_011544388.1 1 360
XM_017028528.1 XP_016884017.1 1 360
Gene Symbol: LINC00319
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026960.1 1 360
GenBank mRNA AK128444.1 - 1 360
BC128187.1 - 1 298
BC128188.1 - 1 298

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline