This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXW12 SIRNA Assays ›
Gene Symbol:
FBXW12
Gene Name:
F-box and WD repeat domain containing 12
Gene Aliases:
FBW12, FBXO12, FBXO35
Chromosome Location:
Chr.3: 48372219 - 48394830 on Build GRCh38
UniGene ID:
Hs.288793
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS138688
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXW12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159927.1 NP_001153399.1 7, 8 977
NM_001159929.1 NP_001153401.1 7, 8 986
NM_207102.2 NP_996985.2 8, 9 1160
XM_017006224.1 XP_016861713.1 7, 8 1173
GenBank mRNA AK097594.1 - 2, 3 278
AY247969.1 - 8, 9 1160
BC101303.1 - 7, 8 1333 40024696
BC101304.1 - 7, 8 921 40024699
BC101305.1 - 7, 8 930 40024702
BC101306.1 - 5, 6 668

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline