siRNA Details
| Gene Symbol: SPOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001664.2 | NP_001001664.1 | 9, 10 | 1371 | ||
| XM_005263655.4 | XP_005263712.1 | 10, 11 | 1334 | |||
| XM_005263656.4 | XP_005263713.1 | 10, 11 | 1263 | |||
| XM_005263657.4 | XP_005263714.1 | 9, 10 | 1176 | |||
| GenBank mRNA | AK293850.1 | - | 3, 4 | 286 | ||
| BC071613.1 | - | 9, 10 | 1350 | 5275316 | ||

SPOPL
Genomic Map