This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF530 SIRNA Assays ›
Gene Symbol:
ZNF530
Gene Name:
zinc finger protein 530
Gene Aliases:
-
Chromosome Location:
Chr.19: 57599877 - 57612722 on Build GRCh38
UniGene ID:
Hs.662177
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS139459
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF530
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321981.1 NP_001308910.1 4 1280
NM_020880.4 NP_065931.3 3 1228
NR_135923.1 3 1228
XM_011526919.2 XP_011525221.1 3 1223
XM_011526921.2 XP_011525223.1 4 1285
GenBank mRNA AB040941.1 - 1 1879
AK096831.1 - 3 1226
AL832740.1 - 3 1228
BC060865.1 - 3 1228 30343224

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0