siRNA Details
| Gene Symbol: OR6C76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005183.1 | NP_001005183.1 | 1 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6C76| Gene Symbol: OR6C76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005183.1 | NP_001005183.1 | 1 | 882 | ||