This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDK18 SIRNA Assays ›
Gene Symbol:
CDK18
Gene Name:
cyclin dependent kinase 18
Gene Aliases:
PCTAIRE, PCTAIRE3, PCTK3
Chromosome Location:
Chr.1: 205504556 - 205532793 on Build GRCh38
UniGene ID:
Hs.445402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS140849
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDK18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002596.3 NP_002587.2 4 635
NM_212502.2 NP_997667.1 4 553
NM_212503.2 NP_997668.1 4 643
XM_011509602.2 XP_011507904.1 4 693
XM_017001423.1 XP_016856912.1 4 788
GenBank mRNA AB209307.1 - 2 1215
AK293756.1 - 3 917
AK295272.1 - 2 83
AK295654.1 - 3 698
AK296320.1 - 4 601
AK308913.1 - 2 84
AU130437.1 - 4 514
AY353237.1 - 4 468
AY353238.1 - 4 532
AY353239.1 - 4 355
BC011526.1 - 4 488 3855304
BT007299.1 - 3 285
BU165355.1 - 555
DC381358.1 - 4 556

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cell cycle
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-tqh7s:80/100.66.75.204:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0