siRNA Details
| Gene Symbol: ARSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001669.3 | NP_001660.2 | 2 | 217 | ||
| NM_009589.3 | NP_033667.2 | 2 | 217 | |||
| XM_005274514.2 | XP_005274571.1 | 2 | 187 | |||
| XM_005274515.2 | XP_005274572.1 | 2 | 187 | |||
| GenBank mRNA | AF160499.1 | - | 2 | 117 | ||
| AK314479.1 | - | 2 | 181 | |||
| BC020229.1 | - | 145 | ||||
| BC080179.1 | - | 2 | 120 | |||
| DA869167.1 | - | 2 | 217 | |||
| X83572.1 | - | 2 | 193 | |||

ARSD
Genomic Map