This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDSN SIRNA Assays ›
Gene Symbol:
CDSN PSORS1C1
Gene Name:
corneodesmosin
psoriasis susceptibility 1 candidate 1
Gene Aliases:
HTSS, HTSS1, HYPT2, PSS, PSS1, S
C6orf16, SEEK1
Chromosome Location:
Chr.6: 31115088 - 31120475 on Build GRCh38
Chromosome Location:
Chr.6: 31114831 - 31140092 on Build GRCh38
UniGene ID:
Hs.310958
Hs.310958
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS141537
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDSN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001264.4 NP_001255.3 2 960
GenBank mRNA AF030130.1 - 2 918
AK292636.1 - 2 928
BC031993.1 - 2 908 4747362
L20815.1 - 2 918
Gene Symbol: PSORS1C1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX640954.1 - 2 1714

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
epidermis development
single organismal cell-cell adhesion
keratinocyte differentiation
skin morphogenesis
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline