This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GCDH SIRNA Assays ›
Gene Symbol:
GCDH
Gene Name:
glutaryl-CoA dehydrogenase
Gene Aliases:
ACAD5, GCD
Chromosome Location:
Chr.19: 12891129 - 12915345 on Build GRCh38
UniGene ID:
Hs.532699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS142163
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GCDH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000159.3 NP_000150.1 6 552
NM_013976.3 NP_039663.1 6 552
NR_102316.1 6 607
NR_102317.1 5 860
XM_006722721.2 XP_006722784.1 6 521
XM_011527899.2 XP_011526201.1 6 521
XM_011527900.2 XP_011526202.1 6 521
XM_017026580.1 XP_016882069.1 6 521
GenBank mRNA AK098370.1 - 6 521
AK290407.1 - 6 521
AK296446.1 - 6 570
AK298772.1 - 4 422
AK300841.1 - 5 809
AK309892.1 - 5 809
AU135567.1 - 6 521
BC002579.2 - 6 505 3142978
BT006706.1 - 5 444
U69141.1 - 5 480

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

lysine catabolic process
tryptophan metabolic process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
fatty-acyl-CoA biosynthetic process
lipid homeostasis
fatty-acyl-CoA binding
glutaryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline