This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CXCL1 SIRNA Assays ›
Gene Symbol:
CXCL1
Gene Name:
C-X-C motif chemokine ligand 1
Gene Aliases:
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Chromosome Location:
Chr.4: 73869392 - 73871302 on Build GRCh38
UniGene ID:
Hs.708652
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS142304
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CXCL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001511.3 NP_001502.1 3 318
NR_046035.1 3 341
GenBank mRNA BC011976.1 - 3 306 3856841
BG121608.1 - 3 325
BM554448.1 - 3 331
BT006880.1 - 3 239
CB112830.1 - 3 319
J03561.1 - 3 292
X12510.1 - 3 256

Back To Top

More Information


Panther Classification:

Molecular Function -

chemokine

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
nervous system development
cell proliferation
negative regulation of cell proliferation
actin cytoskeleton organization
response to lipopolysaccharide
intracellular signal transduction
positive regulation of catalytic activity
cell chemotaxis
chemokine-mediated signaling pathway
positive regulation of neutrophil chemotaxis
receptor binding
chemokine activity
enzyme activator activity
growth factor activity
CXCR chemokine receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline