siRNA Details
| Gene Symbol: KIAA0825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145678.1 | NP_001139150.1 | 18, 19 | 3707 | ||
| XM_006714588.2 | XP_006714651.1 | 18, 19 | 3722 | |||
| XM_011543324.2 | XP_011541626.1 | 18, 19 | 3722 | |||
| XM_011543325.2 | XP_011541627.1 | 19, 20 | 3788 | |||
| XM_011543327.2 | XP_011541629.1 | 18, 19 | 3638 | |||
| XM_011543328.2 | XP_011541630.1 | 19, 20 | 3843 | |||
| XM_011543329.2 | XP_011541631.1 | 17, 18 | 3558 | |||
| XM_011543330.2 | XP_011541632.1 | 18, 19 | 3722 | |||
| XM_011543331.1 | XP_011541633.1 | 18, 19 | 3722 | |||
| XM_011543332.1 | XP_011541634.1 | 18, 19 | 3722 | |||
| XM_011543333.2 | XP_011541635.1 | 18, 19 | 3722 | |||
| XM_011543334.2 | XP_011541636.1 | 18, 19 | 3722 | |||
| XM_011543335.2 | XP_011541637.1 | 15, 16 | 3255 | |||
| XM_017009373.1 | XP_016864862.1 | 18, 19 | 3722 | |||
| XM_017009374.1 | XP_016864863.1 | 18, 19 | 3722 | |||
| XM_017009375.1 | XP_016864864.1 | 19, 20 | 3787 | |||
| GenBank mRNA | AK130941.1 | - | 2, 3 | 332 | ||

KIAA0825
Genomic Map