siRNA Details
| Gene Symbol: RRP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015056.2 | NP_055871.1 | 7 | 678 | ||
| GenBank mRNA | AK056263.1 | - | 7 | 641 | ||
| BC014005.2 | - | 7 | 650 | |||
| BC028386.2 | - | 7 | 683 | 4821672 | ||
| D80001.1 | - | 7 | 576 | |||

RRP1B
Genomic Map