This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PACS2 SIRNA Assays ›
Gene Symbol:
PACS2
Gene Name:
phosphofurin acidic cluster sorting protein 2
Gene Aliases:
PACS-2, PACS1L
Chromosome Location:
Chr.14: 105300811 - 105398147 on Build GRCh38
UniGene ID:
Hs.525626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS146278
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PACS2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100913.2 NP_001094383.2 5 1038
NM_001243127.2 NP_001230056.1 5 584
NM_015197.3 NP_056012.2 5 1038
XM_006720090.1 XP_006720153.1 5 703
XM_006720091.1 XP_006720154.1 5 703
XM_006720092.2 XP_006720155.1 5 703
XM_017021105.1 XP_016876594.1 5 703
XM_017021106.1 XP_016876595.1 5 703
XM_017021107.1 XP_016876596.1 5 703
XM_017021108.1 XP_016876597.1 5 703
XM_017021109.1 XP_016876598.1 5 703
XM_017021110.1 XP_016876599.1 5 703
XM_017021111.1 XP_016876600.1 5 584
XM_017021112.1 XP_016876601.1 5 471
GenBank mRNA AB011174.1 - 5 709
AY320284.2 - 5 1037
BC010663.1 - 2 145
BC050351.1 - 5 576 5264871
BC065220.1 - 5 651 5732786
BC131591.1 - 5 562

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
apoptotic process
viral process
protein localization to pre-autophagosomal structure
protein targeting to plasma membrane
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-tzrzr:80/100.66.76.99:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline