This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GTPBP8 SIRNA Assays ›
Gene Symbol:
GTPBP8
Gene Name:
GTP binding protein 8 (putative)
Gene Aliases:
HSPC135
Chromosome Location:
Chr.3: 112989275 - 113001966 on Build GRCh38
UniGene ID:
Hs.127496
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS147051
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GTPBP8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014170.2 NP_054889.2 6 887
NM_138485.1 NP_612494.1 5 788
GenBank mRNA AF161484.1 - 6 926
AK093419.1 - 6 1481
AK289611.1 - 5 750
AK292651.1 - 6 906
BC010137.2 - 4 623
BC037163.1 - 6 890 4620206
BX354218.2 - 5 756
CA429378.1 - 1 316

Back To Top

More Information


Gene Ontology Categories:

Process(es)

GTPase activity
GTP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline