This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSD17B14 SIRNA Assays ›
Gene Symbol:
HSD17B14
Gene Name:
hydroxysteroid 17-beta dehydrogenase 14
Gene Aliases:
DHRS10, SDR47C1, retSDR3
Chromosome Location:
Chr.19: 48813014 - 48836688 on Build GRCh38
UniGene ID:
Hs.18788
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS147320
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HSD17B14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016246.2 NP_057330.2 6 725
XM_005258969.3 XP_005259026.1 6 734
XM_005258970.3 XP_005259027.1 5 644
GenBank mRNA AF126781.1 - 6 571
AK307437.1 - 6 534
AY358430.1 - 6 564
BC006283.2 - 6 518 4026397
BC006294.1 - 6 526 4025809

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

estrogen biosynthetic process
steroid catabolic process
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
protein binding
identical protein binding
testosterone 17-beta-dehydrogenase (NADP+) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0