siRNA Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142305.1 | NP_001135777.1 | 3 | 1111 | ||
| NM_016643.3 | NP_057727.2 | 3 | 1124 | |||
| GenBank mRNA | AF242768.1 | - | 2 | 753 | ||
| AK313930.1 | - | 3 | 1231 | |||
| BC011870.2 | - | 2 | 873 | 2960099 | ||
| BC026192.2 | - | 3 | 910 | 4801104 | ||

ZNF771
Genomic Map