siRNA Details
| Gene Symbol: C21orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021254.2 | NP_067077.1 | 1 | 470 | ||
| NR_036552.1 | 1 | 465 | ||||
| GenBank mRNA | AF282851.1 | - | 1 | 218 | ||
| AK000474.1 | - | 1 | 465 | |||
| AK055328.1 | - | 2 | 407 | |||
| AK097566.1 | - | 1371 | ||||
| AK098577.1 | - | 1 | 470 | |||
| AK223318.1 | - | 1 | 448 | |||
| AK225697.1 | - | 1 | 448 | |||
| AK300970.1 | - | 1 | 448 | |||
| BC000709.2 | - | 1 | 223 | 3504049 | ||
| FJ649644.1 | - | 1 | 1398 | |||

C21orf59
Genomic Map