This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMIM8 SIRNA Assays ›
Gene Symbol:
SMIM8
Gene Name:
small integral membrane protein 8
Gene Aliases:
C6orf162, dJ102H19.2
Chromosome Location:
Chr.6: 87322588 - 87342328 on Build GRCh38
UniGene ID:
Hs.70769
Species:
Human
siRNA ID HSS148406
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMIM8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042493.2 NP_001035958.1 4 324
NM_020425.5 NP_065158.3 3 303
GenBank mRNA AF311339.1 - 3 345
AK311963.1 - 4 295
AL050201.1 - 2 270
BC070260.1 - 3 285 6452586
BG534658.1 - 3 304
DA293899.1 - 4 295

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-7vrs5:80/100.66.78.233:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0