siRNA Details
| Gene Symbol: LINC00029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028295.1 | 4 | 1365 | |||
| GenBank mRNA | AA609342.1 | - | 1 | 383 | ||
| AL117533.1 | - | 3 | 1247 | |||
| BC069708.1 | - | 4 | 731 | |||
| BC134343.1 | - | 2 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00029| Gene Symbol: LINC00029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028295.1 | 4 | 1365 | |||
| GenBank mRNA | AA609342.1 | - | 1 | 383 | ||
| AL117533.1 | - | 3 | 1247 | |||
| BC069708.1 | - | 4 | 731 | |||
| BC134343.1 | - | 2 | 415 | |||