siRNA Details
| Gene Symbol: BMP8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181809.3 | NP_861525.2 | 7 | 3883 | ||
| GenBank mRNA | AK024413.1 | - | 1 | 296 | ||
| AK127199.1 | - | 1 | 1468 | |||
| BC093743.1 | - | 1 | 59 | |||
| BC112044.1 | - | 1 | 59 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BMP8A| Gene Symbol: BMP8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181809.3 | NP_861525.2 | 7 | 3883 | ||
| GenBank mRNA | AK024413.1 | - | 1 | 296 | ||
| AK127199.1 | - | 1 | 1468 | |||
| BC093743.1 | - | 1 | 59 | |||
| BC112044.1 | - | 1 | 59 | |||