This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF484 SIRNA Assays ›
Gene Symbol:
ZNF484
Gene Name:
zinc finger protein 484
Gene Aliases:
BA526D8.4
Chromosome Location:
Chr.9: 92834262 - 92878060 on Build GRCh38
UniGene ID:
Hs.668378
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS149700
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF484
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007101.2 NP_001007102.1 4 426
NM_001261458.1 NP_001248387.1 4 496
NM_001261459.1 NP_001248388.1 6 567
NM_001261460.1 NP_001248389.1 6 497
NM_031486.2 NP_113674.1 5 471
XM_006717299.3 XP_006717362.1 5 492
XM_011519073.2 XP_011517375.1 6 608
XM_011519074.2 XP_011517376.1 5 381
XM_011519075.2 XP_011517377.1 6 537
XM_017015178.1 XP_016870667.1 7 563
XM_017015179.1 XP_016870668.1 7 411
XM_017015180.1 XP_016870669.1 5 341
XM_017015181.1 XP_016870670.1 7 575
GenBank mRNA AK091203.1 - 4 376
AK292997.1 - 7 555
AK299274.1 - 4 394
AK309617.1 - 5 471
AL833202.1 - 5 441
BC112394.1 - 4 321 40008083
BC112396.1 - 4 321 40008084
DC310695.1 - 5 471
DC418802.1 - 6 498

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-m6gg6:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0