This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF428 SIRNA Assays ›
Gene Symbol:
ZNF428
Gene Name:
zinc finger protein 428
Gene Aliases:
C19orf37, Zfp428
Chromosome Location:
Chr.19: 43607224 - 43619862 on Build GRCh38
UniGene ID:
Hs.99093
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS151404
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF428
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182498.3 NP_872304.2 3 525
GenBank mRNA AY257197.1 - 3 265
BC015950.2 - 3 244 3908182
BC045799.1 - 3 268 4813427
BE901751.1 - 3 513
BG121932.1 - 3 296
CN355560.1 - 3 532

Back To Top

More Information


Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0