siRNA Details
| Gene Symbol: FAM199X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207318.3 | NP_997201.1 | 6 | 1396 | ||
| XM_005262079.2 | XP_005262136.1 | 6 | 1264 | |||
| XM_017029270.1 | XP_016884759.1 | 6 | 965 | |||
| GenBank mRNA | AK091221.1 | - | 7 | 1243 | ||
| AK307312.1 | - | 3 | 193 | |||
| BC016683.1 | - | 4 | 487 | |||
| BC017665.1 | - | 6 | 1251 | 3903800 | ||
| BC035886.1 | - | 5 | 787 | |||
| BC057847.1 | - | 6 | 1396 | 30344004 | ||

FAM199X
Genomic Map