siRNA Details
| Gene Symbol: LINC01465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121682.1 | 1 | 120 | |||
| GenBank mRNA | AK098456.1 | - | 1 | 121 | ||
| BC136288.1 | - | 1 | 114 | 9020275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01465| Gene Symbol: LINC01465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121682.1 | 1 | 120 | |||
| GenBank mRNA | AK098456.1 | - | 1 | 121 | ||
| BC136288.1 | - | 1 | 114 | 9020275 | ||