siRNA Details
| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034145.1 | 2, 3 | 267 | |||
| GenBank mRNA | AK092009.1 | - | 2, 3 | 266 | ||
| DB255288.1 | - | 2, 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00670| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034145.1 | 2, 3 | 267 | |||
| GenBank mRNA | AK092009.1 | - | 2, 3 | 266 | ||
| DB255288.1 | - | 2, 3 | 268 | |||