siRNA Details
| Gene Symbol: LINC01548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024102.1 | 2 | 145 | |||
| GenBank mRNA | BC105115.1 | - | 2 | 145 | ||
| BC105117.1 | - | 2 | 145 | |||
| BX116511.1 | - | 2 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01548| Gene Symbol: LINC01548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024102.1 | 2 | 145 | |||
| GenBank mRNA | BC105115.1 | - | 2 | 145 | ||
| BC105117.1 | - | 2 | 145 | |||
| BX116511.1 | - | 2 | 98 | |||