This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMCO2 SIRNA Assays ›
Gene Symbol:
SMCO2
Gene Name:
single-pass membrane protein with coiled-coil domains 2
Gene Aliases:
C12orf70
Chromosome Location:
Chr.12: 27446482 - 27506911 on Build GRCh38
UniGene ID:
Hs.282121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS155814
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMCO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145010.1 NP_001138482.1 9 1185
XM_011520634.2 XP_011518936.1 9 1363
XM_011520635.2 XP_011518937.1 7 862
XM_017019254.1 XP_016874743.1 7 1119
XM_017019255.1 XP_016874744.1 6 969

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8mgvv:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0