siRNA Details
| Gene Symbol: SPATA31D5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026851.1 | 4 | 372 | |||
| GenBank mRNA | AK125938.1 | - | 4 | 256 | ||
| AK128229.1 | - | 3 | 959 | |||
| BC104780.1 | - | 4 | 292 | |||
| BC104782.1 | - | 4 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA31D5P| Gene Symbol: SPATA31D5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026851.1 | 4 | 372 | |||
| GenBank mRNA | AK125938.1 | - | 4 | 256 | ||
| AK128229.1 | - | 3 | 959 | |||
| BC104780.1 | - | 4 | 292 | |||
| BC104782.1 | - | 4 | 292 | |||