This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM227A SIRNA Assays ›
Gene Symbol:
FAM227A
Gene Name:
family with sequence similarity 227 member A
Gene Aliases:
-
Chromosome Location:
Chr.22: 38578120 - 38656629 on Build GRCh38
UniGene ID:
Hs.542707
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS157825
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM227A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013647.1 NP_001013669.1 15 2078
NM_001291030.1 NP_001277959.1 15 2118
XM_006724291.2 XP_006724354.2 15 2090
XM_006724292.2 XP_006724355.2 15 2078
XM_006724293.2 XP_006724356.1 15 2090
XM_006724294.2 XP_006724357.2 15 2130
XM_006724295.2 XP_006724358.2 15 2118
XM_011530308.2 XP_011528610.1 15 1795
XM_011530309.2 XP_011528611.1 15 2090
XM_011530310.1 XP_011528612.1 14 1985
XM_011530311.1 XP_011528613.1 14 1967
XM_011530312.1 XP_011528614.1 13 1367
XM_011530313.2 XP_011528615.1 13 1319
XM_011530314.1 XP_011528616.1 14 2020
GenBank mRNA AK301819.1 - 15 1792

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline